Documentos clave
Z164313
Supelco
Black plastic caps
Solid top, PTFE-faced rubber liner, thread for 20-400, pkg of 100 ea
About This Item
Código UNSPSC:
41122001
NACRES:
NB.11
Materiales
PTFE-faced rubber liner
black phenolic cap
Características
closure type black phenolic solid top cap
envase
pkg of 100 ea
Adaptador
thread for 20-400
Descripción general
These autoclavable caps are designed to ensure the ultimate in product safety.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)