Resinas para cromatografía de afinidad
La cromatografía de afinidad separa mezclas complejas mediante interacciones bioquímicas muy selectivas. Esta tecnología de purificación muy utilizada separa con eficacia una molécula diana de las impurezas. Nuestra gama de resinas de afinidad está diseñada para una purificación rentable de gran rendimiento, desde el desarrollo de procesos hasta la fabricación a gran escala.
Productos
Recursos relacionados
- Tool: mAbsizer™ Calculator
mAb Expression, Purification, and Formulation
- PDF: Biochromatography Portfolio Guide
Bulk Resin, Membranes and Prepacked Columns
- PDF: Chromatography Purification Selection Guide
Chromatography product selection guide.
- Data Sheet: Eshmuno® A Resin
Eshmuno® A media is a rigid, high-capacity, acid and alkalineresistant Protein A affinity chromatography media for the purification of Fc-containing proteins, including but not limited to monoclonal antibodies.
- Data Sheet: Eshmuno® P anti-A and anti-B Resins
High performance, acid and alkaline resistant affinity chromatography resins designed for the removal of anti-A and anti-B antibodies from plasma-derived immunoglobulin (Ig).
- Data Sheet: Fractogel®Chromatography Resin
Fractogel® resins consist of synthetic methacrylate based polymeric beads providing excellent pressure stability resulting in high flow rates. Depending on your purification application, we can offer resin sized M-type beads with a particle size of 40–90 μm and small S-type beads with a particle size in the range of 20–40 μm.
- Application Note: Using Ethylene Glycol Emprove® Evolve to increase Eshmuno®
Sodium hydroxide (NaOH) is a standard cleaning agent for protein A chromatography resins. Typically, a concentration of 0.1 mol/L NaOH is used. The inclusion of 20% ethylene glycol in a 0.1 mol/L NaOH cleaning solution increases the lifetime of Eshmuno® A columns significantly.
- Application Note: Detection and Quantification of Leached Protein A from Eshmuno® A Resin Using Cygnus F400 Protein A ELISA Kit
Cygnus Technologies performed the initial feasibility testing of the Eshmuno® A Protein A ligand in the Cygnus Protein A ELISA Kit (Cat# F400). Upon successful completion of the feasibility testing, Cygnus proceeded with quantification of the assay for use in in-process and drug substance samples using the Eshmuno® A Protein A resin.
- User Guide: MiniChrom® Pre-Packed Columns
The MiniChrom® columns are prepacked chromatography columns designed for laboratory-scale process development evaluations. When used for biomolecule separations, the results achieved with MiniChrom® columns are typically similar to those obtained using traditional laboratory-scale columns from different vendors.
Resinas de afinidad de proteína A
Eshmuno® A es una resina de gran capacidad para la purificación de proteínas que contienen el fragmento Fc, como los anticuerpos monoclonales (AcM). Esta resina puede desinfectarse en condiciones ácidas y alcalinas.
Resinas de afinidad para purificación del plasma
Las resinas Eshmuno® P anti-A y anti-B eliminan con eficacia los anticuerpos isoaglutininas anti-A y anti-B de las inmunoglobulinas (Ig) plasmáticas, que se han asociado a un mayor riesgo de hemólisis en el paciente.
Resinas de afinidad para purificación de proteínas
La resina Fractogel® de quelato metálico ofrece una separación eficiente y rentable de proteínas y péptidos, especialmente proteínas con restos de histidina, cisteína y triptófano expuestos, incluidas las proteínas etiquetadas con histidina.
Soporte para calificación, evaluación de riesgos y optimización de procesos
Como complemento a nuestra cartera de productos, el programa Emprove® proporciona tres tipos de expedientes para dar soporte a las diferentes etapas de las operaciones de desarrollo y fabricación, como la calificación, la evaluación de riesgos y la optimización de los procesos. Los expedientes concentran datos completos de pruebas específicas del producto, declaraciones de calidad e información normativa en un formato fácilmente disponible para simplificar sus requisitos de cumplimiento.