Documentos clave
26213
Jeringa Hamilton®, 1000 gama GASTIGHT®, cierre Luer de PTFE
1050TLL, Kel-F CTFE Luer lock (with slots), volume 50 mL, needle size (not included)
About This Item
UNSPSC Code:
41122001
eCl@ss:
32070500
En este momento no podemos mostrarle ni los precios ni la disponibilidad
material
Kel-F CTFE Luer lock (with slots)
needle size
(not included)
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 85027
needle L
, n/a
volume
50 mL
fitting
luer lock (without slots)
compatibility
for use with Hamilton 1050TLL (PTFE Luer lock)
Legal Information
GASTIGHT is a registered trademark of Hamilton Co.
Hamilton is a registered trademark of Hamilton Co.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)