Documentos clave
Z275263
Sigma-Aldrich
Cannon-Fenske viscometer tube
size 25
Seleccione un Tamaño
1 EA
$292.200
$292.200
Fecha estimada de envío16 de abril de 2025
Seleccione un Tamaño
Cambiar Vistas
1 EA
$292.200
About This Item
UNSPSC Code:
41111700
NACRES:
NB.61
$292.200
Fecha estimada de envío16 de abril de 2025
size
25
description
constant ~ 0.002 cSt/s
manufacturer/tradename
Ace Glass 798833
viscosity measuring range
0.5-2 cSt
General description
Obtain kinematic viscosities using tubes designed for ASTM Method D445 and D2515. Viscosities can be measured with a precision of ±0.2% quickly and easily.
Requires approximately 7 mL of sample. Certificate of Calibration supplied.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)