Merck
Tutte le immagini(6)
Documenti fondamentali
T4662
Tweezers
Style #5, Fine needle-sharp, Inox alloy (not anti-magnetic)
Sinonimo/i:
fine forceps, fine tweezers, needle tweezers
Scegli un formato
1 EA
CHF 100.00
CHF 100.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 EA
CHF 100.00
About This Item
Codice UNSPSC:
27112105
NACRES:
MA.01
CHF 100.00
Check Cart for Availability
Materiali
Inox alloy (not anti-magnetic)
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Ted Pella 505
Misura
110 mm
Descrizione generale
Specifically designed for microscopy applications. All are ultra-high precision tools of Swiss manufacture.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 9
1 of 9
Tweezers Style #3, very sharp, PTFE-coated, anti-magnetic stainless steel
T5540
Tweezers
Tweezers Style #3C, very sharp, anti-magnetic stainless steel
T4287
Tweezers
Artis tweezer Style 5-SA
Z742676
Artis tweezer
Stainless steel forceps Fine tip, straight
Z168777
Stainless steel forceps
Stainless steel forceps Specimen, L 6 in.
Z168734
Stainless steel forceps
Tweezers Style #3C, very sharp, PTFE-coated, anti-magnetic stainless steel
T5665
Tweezers
Stainless steel forceps Specimen, L 12 in.
Z225622
Stainless steel forceps