Documenti fondamentali
Z162507
Sigma-Aldrich
All-purpose tweezer set
5 tweezers in a vinyl pouch, pkg of 1 set
Scegli un formato
1 SET
CHF 52.60
CHF 52.60
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 SET
CHF 52.60
About This Item
Codice UNSPSC:
41100000
NACRES:
NB.35
CHF 52.60
Check Cart for Availability
Confezionamento
pkg of 1 set
Descrizione generale
Contains 5 tweezers in a vinyl pouch: 41/2 in. dissecting tweezer, 43/4 in. stamp tweezer, 6 in. retrieving forceps, 6 in. solder utility tweezer, and 61/2 in. self-closing tweezer.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 9
1 of 9
Stainless steel forceps Medium tip, straight
Z168750
Stainless steel forceps
Tweezers Style #3C, very sharp, anti-magnetic stainless steel
T4287
Tweezers
Disposable forceps dark blue, L 110 mm, straight tip
Z708356
Disposable forceps
Artis Tweezer Style OO-SA
Z742670
Artis Tweezer
Titanium tweezers Style 2A
Z680214
Titanium tweezers
Tweezers Style #3C, very sharp, PTFE-coated, anti-magnetic stainless steel
T5665
Tweezers
Stainless steel forceps Fine tip, curved
Z168785
Stainless steel forceps
Tweezers Style #7, sharp, hooked, anti-magnetic stainless steel
T4912
Tweezers