Documenti fondamentali
BAF148480000
Fillpour® Dialysis Tube Funnels
5, 9 and 13.5 mm Diameter Steps, 10 pak of
Scegli un formato
10 EA
CHF 111.00
CHF 111.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
10 EA
CHF 111.00
About This Item
Codice UNSPSC:
41123100
NACRES:
NB.24
CHF 111.00
Check Cart for Availability
Materiali
polypropylene
Produttore/marchio commerciale
Bel-Art F14848-0000
Scienceware®
Confezionamento
10 pak of
Categorie correlate
Descrizione generale
To use, cut dialysis tubing to length and knot one end. Slip the open end onto the tube funnel and secure with included rubber band or Scienceware® silicone rubber loops. A captive snap on cover has a small writing area for easy identification. Steam autoclavable at 121 deg C (250 deg F). 10 per bag.
Supports dialysis tubes on edge of vessel and allows easy filling.
Caratteristiche e vantaggi
  • Durable polypropylene construction
  • Allows convenient handling of dialysis bags
  • Three tube steps fit various sizes of dialysis tubing
  • Hanger has three different notches to fit the edge of any vessel
Note legali
Fillpour is a registered trademark of SP Scienceware, Division of SP
Scienceware is a registered trademark of SP Scienceware, a division of SP
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)