Documenti fondamentali
Z371157
Sigma-Aldrich
Dialysis tubing closures
black, size 50 mm
Scegli un formato
10 EA
CHF 122.00
CHF 122.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
10 EA
CHF 122.00
About This Item
Codice UNSPSC:
41123102
NACRES:
NB.22
CHF 122.00
Check Cart for Availability
Materiali
black
Misura
50 mm
Categorie correlate
Descrizione generale
Clamping closures gently lock over dialysis tubing, providing a leakproof seal without knots. Can be used with cellulose ester, regenerated cellulose, and PVDF dialysis membranes. Made entirely of Nylon 66, closures have an operating temperature range of 0-90°C, and can be sterilized with ethylene oxide, γ-irradiation, ethanol, or formalin.
Different colors can be used to code samples. For best results, select a closure 4-10 mm wider than the flat width of the dialysis tubing.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)