Documenti fondamentali
Z562106
Platinum crucible lid
fits 30 mL crucible
Sinonimo/i:
crucibles
Scegli un formato
1 EA
CHF 827.00
CHF 827.00
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 EA
CHF 827.00
About This Item
Codice UNSPSC:
41122200
NACRES:
NB.43
CHF 827.00
Check Cart for Availability
Produttore/marchio commerciale
Birmingham metals
Peso
6.0 g
Descrizione generale
Platinum labware has excellent resistance to acids such as acetic, hydrochloric, hydrofluoric, hydroiodic, nitric, selenic, and sulfuric, and to oxidation at high temperatures. Crucible lids of platinum can be cleaned by boiling in a suitable solvent.
For the removal of substances which have alloyed with the surface, scour with alumina powder.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)