Documenti
Z562033
Platinum crucible lid
fits 10 mL crucible
Sinonimo/i:
crucibles
About This Item
Codice UNSPSC:
41122200
NACRES:
NB.43
Produttore/marchio commerciale
Birmingham metals
Peso
2.8 g
Descrizione generale
Platinum labware has excellent resistance to acids such as acetic, hydrochloric, hydrofluoric, hydroiodic, nitric, selenic, and sulfuric, and to oxidation at high temperatures. Crucible lids of platinum can be cleaned by boiling in a suitable solvent.
For the removal of substances which have alloyed with the surface, scour with alumina powder.
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 2
1 of 2
Platinum combustion boat capacity 3 mL, with handle
Z685364
Platinum combustion boat
Platinum crucible standard, capacity 25 mL
Z562076
Platinum crucible