Documenti fondamentali
Z116149
Stainless steel 304 syringe needle
gauge 22, L 6 in.
Sinonimo/i:
Steel syringe needle
Scegli un formato
1 EA
CHF 54.40
CHF 54.40
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 EA
CHF 54.40
About This Item
Codice UNSPSC:
42142600
eCl@ss:
32070500
NACRES:
NB.03
CHF 54.40
Check Cart for Availability
Descrizione
Standard Hub with 304 Stainless Steel Tubing
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Cadence Science 4176
Lunghezza
6 in.
Gauge
22
Categorie correlate
Descrizione generale
304 SS needle with chrome-plated brass Luer hub. Needle is designed with a slightly curved, deflecting tip to prevent coring during needle insertion through the septum.
Standard Hub with 304 Stainless Steel Tubing
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
Questions
  1. What does the "304" stand for in the description of the Stainless steel 304 syringe needle?
    1 answer
    1. The "304" in Stainless Steel 304 syringe needle stands for the grade or type of stainless steel that it is made from, as in metallurgy.