Documenti fondamentali
Z231029
Sigma-Aldrich
Chem-FLEX transfer line
tube L 30 in.
Sinonimo/i:
Flexible transfer line
Scegli un formato
1 EA
CHF 93.90
CHF 93.90
Check Cart for Availability
Scegli un formato
Cambia visualizzazione
1 EA
CHF 93.90
About This Item
Codice UNSPSC:
41100000
NACRES:
NB.43
CHF 93.90
Check Cart for Availability
Lunghezza
30 in.
Descrizione generale
Two 12 gauge SS needles (6 and 18 in.) are connected to the CHEM-FLEX 106 tubing with nylon clamps. Liquids contact only PTFE and SS during transfers. For extra security when using large bore transfer needles, fasten septum to bottle or flask opening with a wrap-it tie.
The nylon clamps are replaceable, see Z224170.
Please note: To see all 58 stainless steel 304 syringe needles with non-coring points click here
Note legali
Chem-Flex is a trademark of Sigma-Aldrich Co. LLC
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 6
1 of 6
Contenuto correlato
Packaging options for solvents include glass, PVC coated, Sure/Seal bottles, and aluminum cans, offering diverse dispensing solutions.
Questions
  1. What material is the Chem Flex tubing made out of?
    1 answer
    1. This tubing is comprised of PTFE - Poly(tetrafluoroethylene).