Merck
57548
Statischer Mischer mit geringem Volumen (50-250 μl)
housing, In-line, stainless steel
About This Item
UNSPSC-Code:
41115854
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Materialien
stainless steel
Kompatibilität
for use with ASI low volume mixer cartridge
Allgemeine Beschreibung
Low Volume Mixer Housing (housing, In-Line) is an accessory for use with low volume mixer cartridge (50-250 μL) in ASI Static Mixers, used for HPLC and post-column derivatization. “Inline” refers to a single line going into the mixer housing. It is mainly used when additional mixing is needed in a single flow-path.
Empfohlene Produkte
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)