Merck
Z637117
Deckel für BRAND® UV-Küvetten, PE
round blue lid
Größe auswählen
100 EA
CHF 14.60
CHF 14.60
Check Cart for Availability
Größe auswählen
Ansicht ändern
100 EA
CHF 14.60
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.17
CHF 14.60
Check Cart for Availability
Materialien
round blue lid
Hersteller/Markenname
BRAND
Allgemeine Beschreibung
Runde, dicht schliessende Abdeckungen. Sie ermöglichen die Lagerung der Proben bis zu -20°C.
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Kunden haben sich ebenfalls angesehen
Slide 1 of 1
1 of 1
  1. Which document(s) contains shelf-life or expiration date information for a given product?
    If available for a given product, the recommended re-test date or the expiration date can be found on the Certificate of Analysis.
  2. How do I get lot-specific information or a Certificate of Analysis?
    The lot specific COA document can be found by entering the lot number above under the "Documents" section.
  3. How do I find price and availability?
    There are several ways to find pricing and availability for our products. Once you log onto our website, you will find the price and availability displayed on the product detail page. You can contact any of our Customer Sales and Service offices to receive a quote.  USA customers:  1-800-325-3010 or view local office numbers.
  4. What is the Department of Transportation shipping information for this product?
    Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product. 
  5. With what kind of plastic are the BRAND UV cuvettes made? Are they compatible with organic solvents like DMSO, as well as acids and bases?
    The BRAND (Brandtech Scientific) UV cuvettes are made of a proprietary plastic material that has excellent compatibility with acetone, acetonitrile, ethanol, isopropanol and DMSO (dimethyl sulfoxide), The material also has excellent compatibility with acids (hydrochloric acid, sulfuric acid and trichloroacetic acid solution), and bases (sodium hydroxide solution). The cuvettes are not compatible with hydrocarbons (pentane, hexane, heptane, benzene and toluene) or chlorinated solvents (chloroform, carbon tetrachloride and trichloroethylene).
  6. My question is not addressed here, how can I contact Technical Service for assistance?
    Ask a Scientist here.