Merck
Z328650
BRAND® Magnetrührstäbchen, PTFE, zylindrisch
L 10 mm, bar diam. 3 mm
Synonym(e):
PTFE, Stirring rods
Größe auswählen
10 EA
CHF 54.40
CHF 54.40
Check Cart for Availability
Größe auswählen
Ansicht ändern
10 EA
CHF 54.40
About This Item
UNSPSC-Code:
41103806
eCl@ss:
32100100
NACRES:
NB.72
CHF 54.40
Check Cart for Availability
Materialien
PTFE-coated
cylindrical
Hersteller/Markenname
BRAND 137108
Länge
10 mm
Stabdurchmesser
3 mm
Allgemeine Beschreibung
BRAND® magnetic cylindrical stirring bars are expertly designed for optimal mixing performance, featuring a smooth, round shape that enhances stirring efficiency. Their Alnico V magnetic cores are protected by a robust PTFE (polytetrafluoroethylene) coating, ensuring durability and resistance to chemical damage. These stirring bars are particularly effective for use in large volumes or thick mixtures, making them ideal for a variety of laboratory applications.
Leistungsmerkmale und Vorteile
  • BRAND® magnetic cylindrical stirring bars are round with a smooth surface.
  • The Alnico V magnetic cores are encapsulated with high-grade, crack-resistant PTFE (polytetrafluoroethylene) coating.
  • Ideal for large volumes or highly viscous mixtures.
Sonstige Hinweise
BR137108 (India)
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)