Merck
TMO369647
Nalgene® CryoBox
for 3.6-4.5 mL tubes, cardboard, 81 positions, case of 30 ea
Synonym(e):
cryovial storage box, freezer storage
Größe auswählen
30 EACH
CHF 564.00
CHF 564.00
Check Cart for Availability
Größe auswählen
Ansicht ändern
30 EACH
CHF 564.00
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.12
CHF 564.00
Check Cart for Availability
Materialien
cardboard
polycarbonate
Verpackung
case of 30 ea
Hersteller/Markenname
Nalgene 369647
Allgemeine Beschreibung
For ultra-low temperature storage of cryogenic vials in mechanical or liquid nitrogen freezers. Usable temperature range: −196 to +121 °C. Numbers molded into grid system in box correspond to numbers printed on lid. Lid is keyed to box so markings will always match. Boxes accept writing with markers.
Rechtliche Hinweise
CryoBoxes is a trademark of Thermo Fisher Scientific or its subsidiaries
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)