Merck
TMO369639
Nalgene® CryoBox
for 1.0-1.8 mL tubes, cardboard, 81 positions, case of 48 ea
Synonym(e):
cryovial storage box, freezer storage
Größe auswählen
48 EACH
CHF 828.00
CHF 828.00
Check Cart for Availability
Größe auswählen
Ansicht ändern
48 EACH
CHF 828.00
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.12
CHF 828.00
Check Cart for Availability
Materialien
cardboard
polycarbonate
Verpackung
case of 48 ea
Hersteller/Markenname
Nalgene 369639
Allgemeine Beschreibung
For ultra-low temperature storage of cryogenic vials in mechanical or liquid nitrogen freezers. Usable temperature range: −196 to +121 °C. Numbers molded into grid system in box correspond to numbers printed on lid. Lid is keyed to box so markings will always match. Boxes accept writing with markers.
Rechtliche Hinweise
CryoBoxes is a trademark of Thermo Fisher Scientific or its subsidiaries
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)