Merck
TMO278616
Nunc® 1mL 96 DeepWell(TM) plate Well Cap Mat
chemically resistant, sterile, case of 25 ea
Synonym(e):
plate cover, plate cover mat
Größe auswählen
25 EACH
CHF 509.00
CHF 509.00
Check Cart for Availability
Größe auswählen
Ansicht ändern
25 EACH
CHF 509.00
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.15
CHF 509.00
Check Cart for Availability
Sterilität
sterile
Verpackung
case of 25 ea
Hersteller/Markenname
Nunc 278616
Verwandte Kategorien
Allgemeine Beschreibung
Protect plate contents during short- and long-term storage of chemicals or compounds by using the flexible Thermo Scientific Nunc 96 Well Caps with 96 indentations specifically desgined for the 96 wells in Thermo Scientific Nunc 1.0mL Polystyrene DeepWell Plates (278605 and 278606).
Rechtliche Hinweise
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)