Merck
BR701368
BRAND® cover mat for 96-well plates, deep-well
2 mL low profile, TPE, non-sterile
Synonym(e):
Accessories, Cover mats, Deep-well Plates, Plates, Sample Storage
Größe auswählen
50 EA
CHF 270.00
CHF 270.00
Check Cart for Availability
Größe auswählen
Ansicht ändern
50 EA
CHF 270.00
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.22
CHF 270.00
Check Cart for Availability
Materialien
TPE
Sterilität
non-sterile
Verpackung
pack of 50 ea
Hersteller/Markenname
BRAND 701368
Größe
96 well
Allgemeine Beschreibung
These mats provide a reliable barrier to minimize evaporation and contamination during experiments. Cover mats reduce the maximum volume of wells. Adhesive sealing films can also be used.
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)