Merck
T0818
Pharmed L/S tubing
Größe auswählen
25 FT
CHF 216.00
CHF 216.00
Check Cart for Availability
Größe auswählen
Ansicht ändern
25 FT
CHF 216.00
About This Item
UNSPSC-Code:
41105108
NACRES:
NB.35
CHF 216.00
Check Cart for Availability
Verpackung
pack of 25 ea
ID
0.12 in. (3.1 mm)
Größe
Allgemeine Beschreibung
Tubing for MasterFlex Peristaltic Pumps, PharMed tubing has been specifically formulated for the MasterFlex pump system. Great for tissue and cell work- nontoxic and nonhemolytic. Long service life minimizes risk of fluid exposure; reduces tubing cost and pump downtime.
  • Opaque to UV and visible light to protect light-sensitive fluids
  • Heat sealable, bondable, and formable
  • Extremely low gas permeability
Rechtliche Hinweise
MasterFlex is a trademark of Cole-Parmer Instrument Co.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)