MTCP5200100U
MTC™ Bio ProPette™ LE Adjustable Pipettor
capacity 10-100 μL
Synonym(e):
pipette
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121500
NACRES:
NB.15
Empfohlene Produkte
Leistungsmerkmale
adjustable
manual
no. of channels 1
Verpackung
pack of 1 ea
Hersteller/Markenname
MTC™ Bio P5200-100U
Kapazität
10-100 μL
Verwandte Kategorien
Allgemeine Beschreibung
ProPette LE™ Single-Channel Pipettes provide maximum precision at virtually every conceivable volume. They feature a digital micrometer that is adjustable for precise volume control as well as a mechanical volume lock positioned at the top of the pipette that can be engaged or disengaged with one hand. These pipettors are lightweight and finely balanced for maximum ergonomics.
ProPette™ LE, 10 to 100µL
Leistungsmerkmale und Vorteile
- Adjustable with Roto-Lock
- Light weight with reduced forces
- Full volume range (0.1µL-10mL)
- Superior accuracy and reproducibility
- Outstanding user comfort
Rechtliche Hinweise
MTC is a trademark of MTC Bio
ProPette is a trademark of MTC Bio
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)