Merck
GBL705002
Grace Bio-Labs ONCYTE® SuperNOVA nitrocellulose film slides
3-18 mm x 16 mm NC pads per slide, plain glass
Synonym(e):
nitrocellulose slides, protein microarrays
Größe auswählen
20 EA
CHF 769.00
CHF 769.00
Check Cart for Availability
Größe auswählen
Ansicht ändern
20 EA
CHF 769.00
About This Item
UNSPSC-Code:
41122600
NACRES:
NB.15
CHF 769.00
Check Cart for Availability
Materialien
nitrocellulose (on glass)
Verpackung
pkg of 20 ea
Hersteller/Markenname
Grace Bio-Labs 705002
Außenmaß
25 mm × 75 mm
Bindungsart
high binding surface
Allgemeine Beschreibung
Fluorescence detection is a preferred method for many quantitative protein microarray assays in research and diagnostic applications due to its excellent sensitivity and potential for multiplexing. Use of multiplexed fluorescence-based assays on porous nitrocellulose though has been hampered by high backgrounds at shorter excitation wavelengths (such as the 532 nm fluorescence channel). ONCYTE® SuperNOVA microporous nitrocellulose film slides increase protein microarray assay sensitivity and dynamic range using fluorescent endpoints at multiple wavelengths. Use of SuperNOVA slides can lower the limit of detection with fluorescent endpoints by 3 – 4 orders of magnitude over competing nitrocellulose film slides.
Fluorescence detection is a preferred method for many quantitative protein microarray assaysin research and diagnostic applications due to its excellent sensitivity and potential for multiplexing. Use of multiplexed fluorescence-based assays on porous nitrocellulose though has been hampered by high backgrounds at shorter excitation wave lengths (such as the 532nm fluorescence channel). These ONCYTE™ SuperNOVA™ microporous nitrocellulose film slides increase protein microarray assay sensitivity and dynamic range using fluorescent end points at multiple wave lengths. Use of SuperNOVA™ slides can lower the limit of detection with fluorescent end points by 3 – 4 orders of magnitude over competing nitrocellulose film slides.
Rechtliche Hinweise
NOVA is a trademark of Grace Bio-Labs, Inc.
Oncyte is a registered trademark of Grace Bio-Labs, Inc.
SuperNOVA is a trademark of Grace Bio-Labs, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)