Merck
GBL705004
Grace Bio-Labs ONCYTE® SuperNOVA nitrocellulose film slides
4-15 mm x 15 mm NC pads per slide, plain glass
Synonym(e):
nitrocellulose slides, protein microarrays
Größe auswählen
20 EA
CHF 768.00
CHF 768.00
Check Cart for Availability
Größe auswählen
Ansicht ändern
20 EA
CHF 768.00
About This Item
UNSPSC-Code:
41122600
NACRES:
NB.15
CHF 768.00
Check Cart for Availability
Materialien
nitrocellulose (on glass)
Verpackung
pkg of 20 ea
Hersteller/Markenname
Grace Bio-Labs 705004
Außenmaß
25 mm × 75 mm
Bindungsart
high binding surface
Allgemeine Beschreibung
Fluorescence detection is a preferred method for many quantitative protein microarray assaysin research and diagnostic applications due to its excellent sensitivity and potential for multiplexing. Use of multiplexed fluorescence-based assays on porous nitrocellulose though has been hampered by high backgrounds at shorter excitation wave lengths (such as the 532nm fluorescence channel). These ONCYTE™ SuperNOVA™ microporous nitrocellulose film slides increase protein microarray assay sensitivity and dynamic range using fluorescent end points at multiple wave lengths. Use of SuperNOVA™ slides can lower the limit of detection with fluorescent end points by 3 – 4 orders of magnitude over competing nitrocellulose film slides.
Rechtliche Hinweise
NOVA is a trademark of Grace Bio-Labs, Inc.
Oncyte is a registered trademark of Grace Bio-Labs, Inc.
SuperNOVA is a trademark of Grace Bio-Labs, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)