Merck
EP5910760000
Adapter for Eppendorf® S-4x Universal Large Rotor
for 500 mL conical tubes, pack of 2 ea
About This Item
UNSPSC-Code:
41103911
NACRES:
NB.82
Leistungsmerkmale
Holds 500 mL tubes
for 500 mL conical tubes
Verpackung
pack of 2 ea
Hersteller/Markenname
Eppendorf® 5910760000
Methode(n)
centrifugation: suitable
Kompatibilität
for use with Universal bucket
Allgemeine Beschreibung
Adapters for various sizes of tubes and bottles for use with Eppendorf S-4x Universal large rotor.
Rechtliche Hinweise
Eppendorf is a registered trademark of Eppendorf AG