Merck
CLS6791
Corning® LSE Benchtop Shaking Incubator with Platform
AC/DC input 230 V AC, Schuko plug
Synonym(e):
magnetic stirrer, magnetic stirrers, stirrers
Größe auswählen
1 EA
CHF 4’300.00
CHF 4’300.00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
CHF 4’300.00
About This Item
UNSPSC-Code:
41104405
NACRES:
NB.73
CHF 4’300.00
Check Cart for Availability
Leistungsmerkmale
fits flask  4 x 1L
Parameter
20-300 rpm shaking frequency
4-65 °C temp. range (Temperature Increments: 0.1 deg. C)
Außenmaß B × T × H
37 cm × 53 cm × 40 cm
Allgemeine Beschreibung
Truly a benchtop style, the lid on the Corning benchtop shaking incubator has a low pivot point which provides a space-saving size sure to fit on the lab bench and under shelves. Each unit includes a software package, which provides the option of saving data to an external computer or monitoring rpm and temperature throughout runs. The maximum capacity is 4 x 1L flasks.
Leistungsmerkmale und Vorteile
  • Outstanding temperature uniformity
  • Large viewing area for undisturbed monitoring of samples
  • Broad temperature range
  • Multiple shelving options
  • Precision control shaking mechanism
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.
LSE is a trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Kunden haben sich ebenfalls angesehen
Slide 1 of 1