Merck
Z759309
Sigma-Aldrich
Joint clip, standard taper
joint: 19, blue polyethylene
Synonym(e):
NS joint clamps, NS joint clips, National standard joint clamps, National standard joint clips
About This Item
UNSPSC-Code:
41122413
NACRES:
NB.35
Materialien
blue polyethylene
Verbindung
joint: 19
Verpackung
pkg of 50 ea
Allgemeine Beschreibung
Color coded, rigid polyethylene joint clips fit standard taper joints. Will not scratch glassware or corrode; withstands temperatures up to 120 °C.