Merck
Z682101
Ace rotary evaporator condenser complete "C" assembly
glass, fits Büchi® models R-200/R-205 and R-114/R-144, includes 1 L receiving flask and clamp
Größe auswählen
1 EA
CHF 1’640.00
CHF 1’640.00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
CHF 1’640.00
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.43
CHF 1’640.00
Check Cart for Availability
Materialien
glass
Hersteller/Markenname
Büchi equivalent, 40642
Verwandte Kategorien
Allgemeine Beschreibung
The "C" condenser assembly consists of the main glass outer trap body with a 35/20 inner spherical joint bottom connection that attaches to the receiver flasks.
Features standard top and side flange that connects directly to the Büchi® rotary evaporator. The assembly also includes the glass cold trap inner body, PTFE sealing ring with Viton® O-rings, PTFE lid and hose barbs with GL threaded caps.
Rechtliche Hinweise
Büchi is a registered trademark of Büchi Labortechnik AG
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)