Merck
Z645192
IKA® A 11.3 hard metal beater
A 11.3 beater, coated with chromium carbide, 1/cs
Größe auswählen
1 EA
CHF 276.00
CHF 276.00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
CHF 276.00
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.74
CHF 276.00
Check Cart for Availability
Materialien
stainless steel
Verpackung
case of 1
pack of 1
Hersteller/Markenname
IKA 2983000
Eignung
suitable for (pulverizing substances with a Mohs hardness up to 9)
Allgemeine Beschreibung
Stainless stell rackets for use with the IKA A11 Basic Analytical Mill, for breaking hard substances up to Mohs hardness 9,
Rechtliche Hinweise
IKA is a registered trademark of IKA-Werke GmbH & Co. KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)