Merck
Z166898
Sigma-Aldrich
Aldrich® Glas-Laborkit für luftempfindliche Chemikalien
joint: ST/NS 14/20 grease-free, threaded
Synonym(e):
Aldrich glassware
Größe auswählen
1 KIT
CHF 1’060.00
CHF 1’060.00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 KIT
CHF 1’060.00
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.43
CHF 1’060.00
Check Cart for Availability
Verbindung
joint: ST/NS14/20 grease-free, threaded
Allgemeine Beschreibung
Glassware with greaseless, threaded 14/20 joints require no clamps. The unique threaded nut with O-ring connectors in this system automatically separate ground-glass joints when unscrewed and reduces the chance of breakage.
This glassware kit is compatible with Aldrich Schlenk-type glassware components which are specifically designed for air and moisture-sensitive manipulations and reactions. Accessorize the basic glassware kit with Schlenk components to extend the range of applications to include drying and evaporation, extractions, filtrations, recrystallizations, washing degassing and more. Supplied in a foam-lined storage case. Thermoplastic screw caps with Viton O-rings usable to 140 °C.
Verlinkung
Finden sie weiterführende Informationen über die Funktionsweise der Schraubverbindungen unter: Threaded Ground Glass Tutorial
Rechtliche Hinweise
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Kit-Komponenten auch einzeln erhältlich
Produkt-Nr.
Beschreibung
SDB
  • Z167460A. Drying tube for Aldrich® student glass kitSDB
  • Z167533B. Vacuum adapter, Joint: ST/NS 14/20SDB
  • Z167444C. Vacuum distillation adapter with threaded 14/20 jointsSDB
  • Z167495D. Round-bottom flask for Aldrich® student glass kitSDB
  • Z167452E. Three-way adapter, 75° angle, female ST/NS 14/20, male ST/NS 14/20 (2)SDB
  • Z167517F. Aldrich® Schlenk-type Vigreux distilling columnSDB
  • Z167436G. Claisen adapter for Aldrich® student glassware kitSDB
  • Z167576H. Aldrich® Schlenk-type hex head stopperSDB
  • Z167509I. Three-neck round-bottom flask for Aldrich® student glass kitSDB
  • Z167479J. Round-bottom flask for Aldrich® student glass kitSDB
  • Z167525K. Aldrich® Schlenk-type Liebig condenserSDB
  • Z167568L. Thermometer adapter, male ST/NS 14/20SDB
  • Z167487M. Round-bottom flask for Aldrich® student glass kitSDB
  • Z122513N. Gas-inlet tube for Aldrich® Glassware kitsSDB
  • Z167541O. Dropping funnel for Aldrich® student glass kitSDB
  • Z166901Screw-cap, bushing and O-ring set for Aldrich® student glass kit, set of 1SDB
Alle anzeigen (16)
Ähnliches Produkt
Produkt-Nr.
Beschreibung
Preisangaben
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)