Merck
Z152994
Sigma-Aldrich
Thermolyne® 45500 temperature controller
AC/DC input 120 V AC
About This Item
UNSPSC-Code:
41112303
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Hersteller/Markenname
Barnstead International CN45515
Allgemeine Beschreibung
Excellent for use with heating mantles and tapes, small water baths, and small heating elements. Utilizes time-on/time-off stepless input control. Built-in cycle and power-on light. One standard, single-duplex outlet at rear of housing. Compensates for ambient temperature and line voltage fluctuations. Complete with 5 ft, 3-wire cord and plug. Max. load current: 15A.
Rechtliche Hinweise
Thermolyne is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)