Merck
Z125679
Sigma-Aldrich
Ace three-neck round-bottom flasks
vertical, capacity 1,000 mL, center joint: ST/NS 29/42, side Joints: ST/NS 24/40 (2)
Synonym(e):
bottom flask, flask round, round flask
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.43
Materialien
borosilicate glass
round bottom flask
three-neck (Vertical)
Verbindung
center joint: ST/NS29/42
side Joints: ST/NS24/40 (2)
Sterilität
non-sterile
Leistungsmerkmale
closure type Standard Taper joint
Verpackung
pack of 1
Hersteller/Markenname
Ace Glass 6944-26
Kapazität
1,000 mL