Merck
SYNF71542L
Synthware four-neck round bottom flask with threaded side arm
capacity 2,000 mL, center joint: ST/NS 45/50, side joint: ST/NS 24/40
Synonym(e):
Synthware F71542L, 4-neck flask, multi-neck reaction flask
Größe auswählen
1 EA
CHF 377.00
CHF 377.00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
CHF 377.00
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.51
CHF 377.00
Check Cart for Availability
Materialien
borosilicate glass flask
four-neck (with threaded side arm)
round bottom
Verbindung
center joint: ST/NS45/50
side joint: ST/NS24/40
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware F71542L
Kapazität
2,000 mL
Allgemeine Beschreibung
Round bottom, four-neck flask with standard taper outer joints and a thermometer joints. Supplied complete with a compression cap and Viton® O-ring. For insertion of thermometers or bleed tubes having an O.D. between 5.0-7.0 mm.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)