Merck
SYNA701420
Synthware straight vacuum adapter with PTFE stopcock
male joint: ST/NS 14/20
Synonym(e):
Synthware A701420, flow control adapter
Größe auswählen
1 EA
CHF 67.20
CHF 67.20
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
CHF 67.20
About This Item
UNSPSC-Code:
41121815
NACRES:
NB.51
CHF 67.20
Check Cart for Availability
Verbindung
male joint: ST/NS14/20
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware A701420
Allgemeine Beschreibung
Used to regulate the flow of gases or liquids into reaction systems. PTFE stopcock has a bore of 2mm. With standard taper inner joint.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)