Merck
SLW1516/10D
Pyrex® Media-Lab Bottles, with cap and pouring ring, with printed trace code
capacity 1000 mL
Synonym(e):
Pyrex, SciLabware 1516/10D
Größe auswählen
10 EA
CHF 184.00
CHF 184.00
Check Cart for Availability
Größe auswählen
Ansicht ändern
10 EA
CHF 184.00
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.17
CHF 184.00
Check Cart for Availability
Materialien
borosilicate glass bottle
polypropylene cap
round bottle
white graduation
Verbindung
Joints threaded neck (GL45)
Sterilität
non-sterile
Leistungsmerkmale
graduations
with cap
with pouring ring
with printed trace code
Verpackung
pack of 10 ea
Hersteller/Markenname
SciLabware 1516/10D
Kapazität
1000 mL
Durchm. × H
101 mm × 225 mm
Allgemeine Beschreibung
  • Bottles complete with polypropylene cap and drip-free pouring ring
  • Complies with ISO 4796
  • Printed with Trace Code for downloadable batch certificate
  • Batch certificate available to download from www.scilabware.com
  • Ideal for mixing and storage of culture medium
  • Sloping shoulders for smooth pouring action
  • White graduations and marking spot
  • Manufactured from temperature and chemically resistant borosilicate glass
  • For use up to 140 deg. C
Rechtliche Hinweise
PYREX is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)