Merck
CLS7036025
Pyrex® VISTA filter flask
capacity 25 mL
Synonym(e):
conical flasks, filtering flask, filtering flasks, flasks erlenmeyer, Corning® 70360
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.43
Materialien
borosilicate glass
flat bottom flask
narrow-mouth
Sterilität
non-sterile
Leistungsmerkmale
closure type ground joint or rubber stopper
Verpackung
case of 12
pack of 6
Hersteller/Markenname
Corning 70360-25
Kapazität
25 mL
Allgemeine Beschreibung
These small filtering flasks are recommended for microchemical use. All flasks have permanent blue enamel marking spots.
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
VISTA is a trademark of Corning, Inc.