Merck
CAD7931
Laboratory pipetting needle with 90° blunt ends
gauge 22, L 1 1/2 in., nickel plated hub
Größe auswählen
12 EA
CHF 361.00
CHF 361.00
Check Cart for Availability
Größe auswählen
Ansicht ändern
12 EA
CHF 361.00
About This Item
UNSPSC-Code:
41122002
NACRES:
NB.03
CHF 361.00
Check Cart for Availability
Materialien
nickel plated hub
Verpackung
pack of 12 ea
Hersteller/Markenname
Cadence Science 7931
Länge
1 1/2 in.
Manometer
22
Allgemeine Beschreibung
Pipetting needles are available in a wide choice of sizes for use in laboratory procedures. All needles are made with type 304 stainless steel cannula.
Catalog numbers CAD7950 to CAD7957 needles are fitted with stainless steel hubs for special applications with corrosive materials or critical procedures. Catalog numbers CAD7930 to CAD7942 needles are fitted with standard nickel plated Luer hubs. These needles are used in conjunction with both manual and automatic volumetric dispensers, dilutors, and pipettors.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)