Merck
295272
Sigma-Aldrich
1,1-Difluor-ethylen
≥99%
Synonym(e):
Vinylidenfluorid
About This Item
Lineare Formel:
H2C=CF2
CAS-Nummer:
Molekulargewicht:
64.03
Beilstein:
1733321
EG-Nummer:
MDL-Nummer:
UNSPSC-Code:
12142100
PubChem Substanz-ID:
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Dampfdichte
2.2 (vs air)
Dampfdruck
518 psi ( 21 °C)
Assay
≥99%
Selbstzündungstemp.
1184 °F
Expl.-Gr.
21.3 %
bp
−83 °C (lit.)
mp (Schmelzpunkt)
−144 °C (lit.)
SMILES String
FC(F)=C
InChI
1S/C2H2F2/c1-2(3)4/h1H2
InChIKey
BQCIDUSAKPWEOX-UHFFFAOYSA-N
Verpackung
Packaged in carbon steel lecture bottles with CGA 110F/180M Al-Si-Bronze needle valve.
Sonstige Hinweise
Rechtliche Hinweise
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Empfehlung
Entlüftungsventil
Produkt-Nr.
Beschreibung
Preisangaben
Schlaucholive
Produkt-Nr.
Beschreibung
Preisangaben
auch häufig zusammen mit diesem Produkt gekauft
Produkt-Nr.
Beschreibung
Preisangaben
Piktogramme
FlameGas cylinder
Signalwort
Danger
H-Sätze
P-Sätze
Gefahreneinstufungen
Flam. Gas 1 - Press. Gas Liquefied gas
Lagerklassenschlüssel
2A - Gases
WGK
WGK 2
Flammpunkt (°F)
Not applicable
Flammpunkt (°C)
Not applicable
Persönliche Schutzausrüstung
Eyeshields, Faceshields, Gloves, multi-purpose combination respirator cartridge (US)
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Kunden haben sich ebenfalls angesehen
Dawei Su et al.
Scientific reports, 2, 924-924 (2012-12-12)
Faceted crystals with exposed highly reactive planes have attracted intensive investigations for applications such as hydrogen production, enhanced catalytic activity, and electrochemical energy storage and conversion. Herein, we report the synthesis of mesoporous NiO crystals with dominantly exposed {110} reactive
Jennifer R Du et al.
Water research, 43(18), 4559-4568 (2009-09-01)
A commercial poly(vinylidene fluoride) flat sheet membrane was modified by surface coating with a dilute poly(vinyl alcohol) (PVA) aqueous solution followed by solid-vapor interfacial crosslinking. The resulting PVA layer increased membrane smoothness and hydrophilicity and resulted in comparable pure water
Alaaeldin Ahmed Hamza et al.
PloS one, 11(11), e0167049-e0167049 (2016-11-24)
Cardiotoxicity is a limiting factor of doxorubicin (DOX)-based anticancer therapy. Due to its beneficial effects, we investigated whether standardized extract of Melissa officinalis (MO) can attenuate doxorubicin-induced cardiotoxicity and can potentiate the efficacy of DOX against human breast cancer cells.
Indro Neil Ghosh et al.
ACS synthetic biology, 5(12), 1519-1534 (2016-07-13)
The optimization of synthetic pathways is a central challenge in metabolic engineering. OptSSeq (Optimization by Selection and Sequencing) is one approach to this challenge. OptSSeq couples selection of optimal enzyme expression levels linked to cell growth rate with high-throughput sequencing
Liyan Wang et al.
FEBS letters, 590(23), 4252-4262 (2016-10-23)
MiR-296 was previously reported to be underexpressed in hepatocellular carcinoma (HCC). However, the clinical value of miR-296 and its function in HCC remain poorly understood. In this study, we found that miR-296 levels are decreased in HCC specimens and cells