Principaux documents
G5778
Sigma-Aldrich
Glass plates for vertical standard gel
16.5 cm × 19 cm
About This Item
Code UNSPSC :
41105312
Nomenclature NACRES :
NB.51
Nom du produit
Glass plates for vertical standard gel, 16.5 cm wide, W × H 16.5 cm × 19 cm
Larg. × H
16.5 cm × 19 cm
Niveau de qualité
Conditionnement
Set includes one rectangular plate with rounded bottom corners and one notched plate.