Principaux documents
Z255858
Sigma-Aldrich
Latex rubber laboratory tubing
I.D. × O.D. 1/4 in. × 5/8 in.
About This Item
Code UNSPSC :
41105108
Nomenclature NACRES :
NB.51
Conditionnement
pkg of 50 feet
D.I. × D.E.
1/4 in. × 5/8 in.
Taille de la paroi
3/16 in.
Description générale
High-purity, seamless, amber tubing for general-purpose use with liquids and gases. Durometer: 35 ±5. Talc-free.