Key Documents
SYNA702440
Synthware straight vacuum adapter with PTFE stopcock
male joint: ST/NS 24/40
Synonyme(s) :
Synthware A702440, flow control adapter
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.51
Joint
male joint: ST/NS24/40
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware A702440
Description générale
Used to regulate the flow of gases or liquids into reaction systems. PTFE stopcock has a bore of 2mm. With standard taper inner joint.
Informations légales
Synthware is a trademark of Kemtech America Inc.