Z219959
Aldrich® hollow pennyhead stopper
joint: ST/NS 34/45
Synonyme(s) :
glass stopper, glass stoppers, stoppers
About This Item
Produits recommandés
Matériaux
ground glass
Joint
joint: ST/NS34/45
Description
closed at bottom
Catégories apparentées
Description générale
Conforms to standard-taper specifications as outlined in ASTM E 675.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC