Principaux documents
Z219932
Sigma-Aldrich
Aldrich® hollow pennyhead stopper
joint: ST/NS 24/40
Synonyme(s) :
glass stopper, glass stoppers, stoppers
About This Item
Code UNSPSC :
41122500
Nomenclature NACRES :
NB.35
Matériaux
ground glass
Joint
joint: ST/NS24/40
Description
closed at bottom
Description générale
Conforms to standard-taper specifications as outlined in ASTM E 675.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC