Principaux documents
DWK258522102
DURAN® Filter funnel
borosilicate glass 3.3, volume 125  mL, porosity 1, autoclavable, pack of 1 ea
Synonyme(s) :
258522102
About This Item
Code UNSPSC :
41104925
Nomenclature NACRES :
NB.43
Matériaux
borosilicate glass 3.3
Caractéristiques
autoclavable
Conditionnement
pack of 1 ea
Fabricant/nom de marque
DURAN® 258522102
Diam. du disque
60 mm
Volume
125  mL
Dimension de pores
1 porosity
Capacité
125 mL capacity
Description générale
One of the most important separation methods in the laboratory environment is filtration, which is used to separate or clean different media. For this working method, filters are required which are selected according to the application with regard to size and porosity. Büchner funnels are cylindrical with an integral sintered disc. Without additional filter paper, only coarse filtration is possible. Büchner funnels can also be used in vacuum filtration. The applied vacuum accelerates the filtration.
Caractéristiques et avantages
• Manufactured from 3.3. expansion borosilicate glass• Funnels have an integral sintered glass filtration disc• Comprehensive range of funnel sizes and sinter porosities (grades) are available• The use of filter paper is not required• Resistant to most corrosive reagents including ammonia and concentrated sulphuric acid• Filter funnels can be used with a filtering flask via a conical rubber seal (GUKO)• Fired-on, highly durable white print
Informations légales
DURAN is a registered trademark of DWK Life Sciences