Principaux documents
T1906
Sigma-Aldrich
Ultramicro pipette tips
Microcapillary, round tip, maximum volume 200 μL, non-sterile
About This Item
Code UNSPSC :
41121600
Nomenclature NACRES :
NB.01
Matériaux
colorless
polypropylene
Stérilité
non-sterile
Caractéristiques
barrier: no
Conditionnement
pack of 200 ea (Trays)
Fabricant/nom de marque
Sigma-Aldrich
Volume maximum
200 μL
Description générale
For loading tube gels and slab gels 0.6 mm or thicker. Mark at 5 μl gives visual confirmation of proper pipet operation. Each tray contains 10 × 20 tips
Low capacity improves accuracy with small sample volumes by reducing dead space.
Autres remarques
Browse all pipette tips