Principaux documents
Z421022
Sigma-Aldrich
Aldrich® swivel McLeod vacuum gauge
McLeod gauge (Oil swivel)
1 EA
1 160,00 $
1 160,00 $
Date d'expédition estimée le21 avril 2025Détails
Changer de vue
1 EA
1 160,00 $
About This Item
Code UNSPSC :
41112406
Nomenclature NACRES :
NB.79
1 160,00 $
Date d'expédition estimée le21 avril 2025Détails
Description
Square root scale: 1 millitorr to 1 torr
linear scale 0 - 1 torr in 0.02 torr div.
Catégories apparentées
Description générale
Made with permanent linear and square root ceramic scales on a white background, high-vacuum O-ring seals, precision bore capillaries, Delrin® plastic and black anodized aluminum parts. Accurate to ±3% of reading or 1mm of scale division, whichever is greater. Gauges include operating instructions but do not include oil or mercury fill. Swivel arrangement permits rigid connection to the vacuum system with the scales vertical at all times.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Delrin is a registered trademark of E. I. du Pont de Nemours and Company