CLS51002L
Pyrex® wide-mouth Erlenmeyer flask
capacity 2,000 mL
Synonyme(s) :
conical flasks, erlenmeyer flasks, flask erlenmeyer, flasks erlenmeyer, Corning® 5100
65,30 $
About This Item
65,30 $
Produits recommandés
Matériaux
borosilicate glass
wide-mouth
Joint
joint stopper13
Stérilité
non-sterile
Caractéristiques
cap: no
subdivision 100
Conditionnement
pack of 1
case of 8
Fabricant/nom de marque
Corning 5100-2L
Plage de mesure du volume
400-1,800 mL
Capacité
2,000 mL
Description générale
- Pyrex® wide-mouth Erlenmeyer flasks are made of borosilicate glass for optical clarity and mechanical strength.
- Flasks have durable white enamel graduations to show approximate capacity and an extra-large marking spot.
- Features a heavy-duty rim.
- Ideal for use as a titration flask.