CLS5100125
Pyrex® wide-mouth Erlenmeyer flask
capacity 125 mL
Synonyme(s) :
conical flasks, erlenmeyer flasks, flask erlenmeyer, flasks erlenmeyer, Corning® 5100
184,00 $
About This Item
184,00 $
Produits recommandés
Matériaux
borosilicate glass
bottom (baffled)
wide-mouth
Joint
joint stopper6
Stérilité
non-sterile
Caractéristiques
cap: no
subdivision 25
Conditionnement
pack of 12
case of 48
Fabricant/nom de marque
Corning 5100-125
Plage de mesure du volume
50-125 mL
Capacité
125 mL
Description générale
- Pyrex® wide-mouth Erlenmeyer flasks are made of borosilicate glass for optical clarity and mechanical strength.
- Flasks have durable white enamel graduations to show approximate capacity and an extra-large marking spot.
- Features a heavy-duty rim.
- Ideal for use as a titration flask.