Principaux documents
CLS53402L
Pyrex® heavy wall graduated filtering flask, with tubulation
12.5 mm O.D. hose-connection, capacity 2,000 mL
Synonyme(s) :
conical flasks, filtering flask, filtering flasks, flasks erlenmeyer, Corning® 5340
1 EA
248,00 $
12 EA
2 290,00 $
248,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 EA
248,00 $
12 EA
2 290,00 $
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.43
248,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
borosilicate glass
flat bottom flask
narrow-mouth
Joint
joint stopper9
Stérilité
non-sterile
Caractéristiques
closure type Rubber stopper
Conditionnement
pack of 1
case of 12
Fabricant/nom de marque
Corning 5340-2L
Plage de mesure du volume
600-2,000 mL
Capacité
2,000 mL
Description générale
These flasks have sidearm tabulations and are blown in special molds designed to give maximum mechanical strength. For convenience, the flasks are graduated to show approximate capacity. The neck finish affords an excellent fit for rubber stoppers.
Informations légales
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.