CLS3953312EC
Pyrex® gas-dispersion tube
pore size 70-220 μm
Synonyme(s) :
Corning® 39533
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
Produits recommandés
Fabricant/nom de marque
Corning 39533-12EC
Dimension de pores
70-220 μm
Description générale
With 12 mm O.D. fritted cylinder, 250 mm L × 8 mm diam. stem. Used to disperse gas in liquids. Size of bubbles depends on porosity. Size permits insertion through small openings.
Informations légales
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.