Principaux documents
Z177482
Scienceware® gas-dispersion tube
polyethylene
4 EA
107,00 $
107,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
4 EA
107,00 $
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.51
107,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
polyethylene
Conditionnement
pack of 4 ea
Fabricant/nom de marque
Bel-Art F13691-0000
Catégories apparentées
Description générale
Linear bell-mouthed fitting has sealed-in PE disc, 70 μm porosity, at large end; 30.5 cm (12 in.) L × 6.35 mm (1/4 in.) I.D. Tube is fitted to the other end, 2.54 cm bell (1 in.) O.D.
Informations légales
Scienceware is a registered trademark of SP Scienceware, a division of SP
Questions
  1. what is the porosity?
    1 answer
    1. Porosity is a measurement of open space in a filter and is not determined for this product. The pore rating of the porous polyethylene disc is considered “COARSE” meaning the pores range in diameter from 90µm to 130µm.